Learn More
Tocris Bioscience™ TM 5441
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Plasminogen activator inhibitor-1 (PAI-1) inhibitor (IC50 = 3.58 - 60.3 μM). Increases apoptosis in human cancer cell lines. Reduces senescence and increases lifespan in mouse models of ageing (kl/kl). Inhibits L-NAME (Cat.No. 0665) induced systolic blood pressure increases, telomere length reduction and cardiac hypertrophy in mice. Renoprotective. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Formula Weight | 450.8 |
| Physical Form | Solid |
| Purity | >98% |
| Chemical Name or Material | 5-Chloro-2-[[2-[2-[[3-(3-furanyl)phenyl]amino]-2-oxoethoxy]acetyl]amino]benzoic acid sodium salt |