Learn More
SI Analytics™ Titroline™ 6000 Titrators
Product Code. 11944729 Shop All SI Analytics Products
Click to view available options
Display Type:
TFT
Unit Size:
Each
Product Code. 11944729
Brand: SI Analytics™ 285220080
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
TFT display allows perfect readout even from the side. The Titroline 6000 Titrator is ideal for food, water, wastewater and environmental applications.
  • Intelligent exchangeable units store all relevant reagent and unit data
  • Automatic wireless recognition of Schott Instruments ID electrodes guarantees accurate calibration and measurements
  • Touch-sensitive glass keypad
  • Three USB and two RS-232 ports
  • High resolution pH/mV electrode and temperature inputs for pH, ISE, redox or photometric titrations
  • Polarizable electrode input for set endpoint applications
  • Available standard methods such as FOS/TAC, alkalinity, total acidity in soft drinks
  • Linear (fixed increment) and dynamic equivalence point titration modes
  • Titrations to pH, mV and μA end point
  • Manual titration mode and routine dosing tasks are also available
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Three USB and two RS-232 ports
Description Titrator
Display Type TFT
For Use With (Application) Food, water, wastewater and environmental applications
Includes TitroLine 6000 basic unit, magnetic stirrer, 20mL exchangeable unit, bottle, adapters, hoses, drip off tube, titration tip; with pH combination electrode and buffer set