Learn More
BRAND™ Titrette™ Adapter Set Interface RS232 to USB
Product Code. 17821252 Shop All Brand Products
Click to view available options
:
For Use With (Application): Connecting Burette to Computer, For Use With (Equipment): Computer or Laptop, Type: USB Adapter
Product Code. 17821252
Brand: BRAND™ 707539
For use in connecting the Titrette burette to a computer.
  • Adapter connects the Titrette™ bottle-top burette (with RS232 interface) to a desktop computer or laptop
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 17.6 cm
For Use With (Equipment) Computer or Laptop
For Use With (Application) Connecting Burette to Computer
Height (Metric) 3.23 cm
Includes RS232 Interface
Type USB Adapter