Learn More
DiversifiedBiotech Titer Tops with 100 Pk.
Description
- Pre-cut feature
- Pierceable polyethylene microplate film for preventing cross-well contamination
- End tabs make film positioning and removal hassle-free
- Titer Tops with 100 Pk.
Specifications
Specifications
| Color | Clear |
| Material | Polyethylene |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| Quantity | 100/Pk. |
| Product Type | Titer Tops Seals |
| Adhesive | Acrylic |
| Pierceable | Yes |