Learn More
Titanium granules, 99.99% (metals basis excluding Na and K)
Product Code. 15466305
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466305 500 g 500g
11314637 50 g 50g
11334637 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15466305 Supplier Thermo Scientific Alfa Aesar Supplier No. 039677.36
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
CAS: 7440-32-6 | Ti | 47.87 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]
Specifications
Quantity 500 g
Odor Odorless
Form Granular
Percent Purity 99.99%
Assay (metals basis excluding Na and K)
Chemical Name or Material Titanium granules