Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Product Code. 12638356 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Pack of 500
Product Code. 12638356
Brand: Epredia™ C1000YL
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Handle small biopsies and fragile specimens with Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures tissues are safely processed as cassettes will not pop open during processing
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Mesh Pore Insert
Color Yellow
Product Type HistoScreen™ Cassette