Learn More
Bel Art™ Tissue Culture Flask Racks
Product Code. 11732016
Change view
Click to view available options
Positions:
5
12
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Unit Size
11732016 5 Each
11722016 12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11732016 Supplier Bel Art™ Supplier No. F189700002
  • ONLINE EXCLUSIVE PRICE
    78.15€
    64.11€ / Pack of 1
    Save 14.04€
    17% Off
View all promotions
Estimated Shipment: 16-12-2026
Racks prevent the domino effect of flasks tipping one over the other.
  • These polypropylene racks hold 25cc, 75cc, 50 to 75mL, or 250 to 260 mL or similar sized tissue culture flasks in a vertical position
  • Rectangular holes in the top and center plates stabilize the flasks preventing tipping
  • Easily organize flasks to improve processing and to avoid errors in collecting used media, media change, washing of cell monolayer, trypsinization and seeding of cells
  • Grips on each end make rack handling easy and allow for safe transport of flasks to the incubator
  • Steam autoclavable at 121°C (250°F)
Specifications
Autoclavable Autoclavable
Color White
Holds Flasks
Material Polypropylene
Positions 5
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) 250 to 260 mL Flasks
Quantity 1 Each
Temperature (Metric) Range 121°C
Dimensions (L x W x H) 29.7 x 12.5 x 13.2 cm
Length (Metric) 29.7 cm
Product Type Flask Rack
Shelves 3 Tiers
Height (Metric) 13.2 cm
Array 5 x 1
Width (Metric) 12.5 cm