Learn More
Epredia™ Tissue Cassette Ii Embedding
Product Code. 12943755 Shop All Epredia Products
Click to view available options
Quantity:
75 cassettes
Unit Size:
Pack of 750
Product Code. 12943755
Brand: Epredia™ B1000729FPK
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Pore style slotted, flat 45' writing angle, acetal polymer fluorescent pink locking lid to open front & back, supplied as 10 tubes of 75 cassettes Epredia Shandon
TRUSTED_SUSTAINABILITY
Specifications
Quantity 75 cassettes