Learn More
Epredia™ Tissue Cassette Ii Embedding
Product Code. 12953755 Shop All Epredia Products
Product Code. 12953755
Brand: Epredia™ B1000729FYW
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Pore style slotted, flat 45' writing angle, acetal polymer fluorescent yellow locking lid to open front & back, supplied as 10 tubes of 75 cassettes Epredia Shandon
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow