Learn More
Sampling Systems™ Tip for PowderThief Sampler 32mm Diameter
Product Code. 15879201 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
20 mL
25 mL
50 mL
100 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15879201 100 mL Each
15156852 20 mL Each
15186852 25 mL Each
15186862 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15879201 Supplier Sampling Systems™ Supplier No. 1030DT100
  • ONLINE EXCLUSIVE PRICE
    128.00€
    123.35€ / Pack of 1
    Save 4.65€
    3% Off
Estimated Shipment: 04-01-2027
Extensively used throughout the pharmaceutical, chemical and food industries around the world. Sampling Systems™ Tip for PowderThief Sampler 32mm Diameter allow a single sampler to be used to accurately sample a range of different volumes.
  • Made from high quality 316L stainless steel
  • Wide range of sample sizes
  • Wide range of sampler lengths
  • Easy to use
  • A range of tips that let a single sampler be used to accurately sample a range of different volumes
Specifications
Product Type Tip
Material Stainless Steel (316L)
Capacity (Metric) 100 mL
For Use With (Application) Powder sampling
Diameter (Metric) 32 mm