Learn More
Sampling Systems™ Tip for PowderThief Sampler 25mm Diameter
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
10 mL
20 mL
25 mL
Unit Size:
Each
Description
- Made from high quality 316L stainless steel
- Wide range of sample sizes
- Wide range of sampler lengths
- Easy to use
- A range of tips that let a single sampler be used to accurately sample a range of different volumes
Specifications
Specifications
| Product Type | Tip |
| Material | Stainless Steel (316L) |
| Capacity (Metric) | 10 mL |
| For Use With (Application) | Powder sampling |
| Diameter (Metric) | 25 mm |