missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiotropium-d3 Bromide (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Tiotropium N-methyl-d3 bromide |
| CAS | 1127226-56-5 |
| Molecular Formula | C19 D3 H19 N O4 S2 . Br |
| Quantity | 0.005 g |
| Synonym | 3-Oxa-9-azoniatricyclo[3.3.1.02,4 ]nonane, 7-[(2-hydroxy-2,2-di-2-thienylacetyl)oxy]-9-methyl-9-(methyl-d3)-, bromide (1:1), (1α,2β,4β,5α,7β)-, Tiotropium-d3 Bromide, Tiotropium-(N-methyl-d3) bromide, (1α,2β,4β,5α,7β)-7-[(2-Hydroxy-2,2-di-2-thienylacetyl)oxy]-9-methyl-9-(methyl-d3)-3-oxa-9-azoniatricyclo[3.3.1.02,4 ]nonane bromide, Tiotropium-D3 (N-methyl-D3) bromide |
| InChI Formula | InChI=1 S/C19H22NO4S2.BrH/c1-20(2)12-9-11(10-13(20)17-16(12)24-17)23-18(21)19(22,14-5-3-7-25-14)15-6-4-8-26-15;/h3-8,11-13,16-17,22 H,9-10H2,1-2H3;1 H/q+1;/p-1/t11-,12-,13+,16-,17+;/i1D3;/t11-,12-,13+,16-,17+,20?; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])[N+]1(C)[C@@H]2 C[C@H](C[C@H]1 [C@@H]3 O[C@H]23)OC(=O)C(O)(c4cccs4)c5cccs5 |
| IUPAC Name | [(1 R,2 R,4 S,5 S)-9-methyl-9-(trideuteriomethyl)-3-oxa-9-azoniatricyclo[3.3.1.02,4 ]nonan-7-yl] 2-hydroxy-2,2-bis(2-thienyl)acetate bromide |
| Molecular Weight (g/mol) | 475.435 |
| Formula Weight | 474.036 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction