Learn More
Lovibond™ Tintometer™ Sulphite No.2 H.R.Tablets
Product Code. 15362005
Click to view available options
Quantity:
100 Tablets
Unit Size:
Pack of 250
Product Code. 15362005
Brand: Lovibond™ 505281
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Sulphite No.2 H.R.Tablets
  • Bottle of 100
Specifications
For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 Tablets
Type Sulphite No.2 H.R.Tablets
compliance-icons
Product Identifier
  • X250 SULFITE NO.2 HR TABLETS .
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide