missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Potassium Sodium Tartrate NAA, 100mL

Product Code. 15342045
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 15342045

Brand: Lovibond™ 466101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers

  • Potassium Sodium Tartrate NAA Solution
  • 100mL

Specifications

Quantity 100 mL
Type Potassium Sodium Tartrate NAA Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.