Learn More
Lovibond™ Tintometer™ Nitrite Acidifying NJP Tablets
Product Code. 15311995
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
Product Code. 15311995
Brand: Lovibond™ 502371
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Nitrite Acidifying NJP Tablets
  • Bottle of 250
Test tablets are manufactured in Germany under tightly controlled conditions on most modern machinery. Maintaining the highest quality standards permits Tintometer to guarantee our tablet reagents for a minimum of 5 years, and some for as long as 10 years.
Specifications
For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Type Nitrite Acidifying NJP Tablets
compliance-icons compliance-icons
Product Identifier
  • X250 NITRITE ACIDIFYING NJP TABLETS .
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H318-Causes serious eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : sodium bisulphate, boric acid