Learn More
Lovibond™ Tintometer™ Nitrite Acidifying NJP Tablets
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Nitrite Acidifying NJP Tablets
- Bottle of 250
Specifications
Specifications
| For Use With (Application) | Photometry Tablet Reagents |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 250 Tablets |
| Type | Nitrite Acidifying NJP Tablets |
Safety and Handling
Product Identifier
- X250 NITRITE ACIDIFYING NJP TABLETS .
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contains : sodium bisulphate, boric acid