Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Sulphite LR Test, 0.5 to 10mg/L
Click to view available options
Range:
0.5 to 10mg/L
Unit Size:
Each
Description
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
- Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
- The Sulphite disc supplied measures from 0.5 to 10mg/L with the following steps: 0.5 / 1 / 1.5 / 2 / 2.5 / 3 / 3.5 / 4 / 4.5 / 5 / 6 / 7 / 8 / 9 / 10.5 / 4.0
Specifications
Specifications
| For Use With (Application) | Water Quality Testing |
| For Use With (Equipment) | Checkit™ Comparator |
| Includes | Carry case with Comparator, Vials, Discs, reagents and method instructions for all tests |
| Range | 0.5 to 10mg/L |
| Type | Sulphite LR Test |