Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Ammonia (Powder Pack) Test, 0 to 0.5mg/L
Product Code. 10105094
Click to view available options
Range:
0 to 0.5mg/L
Unit Size:
Each
Product Code. 10105094
Brand: Lovibond™ 146211
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
  • The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
  • Suitable for both mobile and static water analysis
  • Available with a number of different color scales
  • Provides the basis for a comprehensive easy to to to use analysis system
  • Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
  • Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
  • The Ammonia disc supplied measures from 0 to 0.5mg/L with the following steps:
  • 0, 0.05, 0.1, 0.15, 0.2, 0.25, 0.3, 0.35, 0.4, 0.45, 0.5
Specifications
Format Ammonia Test (Powder Pack)
For Use With (Application) Water Quality Testing
For Use With (Equipment) Checkit™ Comparator
Includes Carry case with Comparator, Vials, Discs, reagents and method instructions for all tests
Range 0 to 0.5mg/L
Type Ammonia Test Disc for Checkit™ Comparator