Learn More
Tin bis(trifluoromethylsulfonyl)imide, Thermo Scientific™
48.75€ - 545.00€
Chemical Identifiers
| CAS | 4600096-08-6 |
|---|---|
| MDL Number | MFCD28100488 |
| Synonym | Tin bis(trifluoromethanesulfonyl)imide; Tin bis(trifluoromethanesulfonimide) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15482867
|
Thermo Scientific Alfa Aesar
047142.MD |
250mg |
48.75€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15462867
|
Thermo Scientific Alfa Aesar
047142.03 |
1g |
139.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15472867
|
Thermo Scientific Alfa Aesar
047142.06 |
5g |
545.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 4600096-08-6 | |
| MFCD28100488 | |
| Tin bis(trifluoromethanesulfonyl)imide; Tin bis(trifluoromethanesulfonimide) |
Specifications
| 4600096-08-6 | |
| MFCD28100488 | |
| Hygroscopic | |
| 678.98 |
| [(CF3SO2)2N]2Sn | |
| UN3261 | |
| Tin bis(trifluoromethanesulfonyl)imide; Tin bis(trifluoromethanesulfonimide) | |
| Tin bis(trifluoromethylsulfonyl)imide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen