missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-5 Rat anti-Mouse, Clone: 339319, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3374-SP
This item is not returnable.
View return policy
Description
TIM-5 Monoclonal specifically detects TIM-5 in Mouse samples. It is validated for Western Blot.Specifications
| TIM-5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_001156829 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 210535 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 339319 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Gm12169 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIM-5 Phe22-Gly161 Accession # NP_001156829 | |
| 25 μg | |
| Primary | |
| Detects mouse TIM-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TIM-1, -2, -3, -4, or -6 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction