Learn More
Oxoid UK Tilmicosin TIL 15ug 5x50 discs
Click to view available options
Quantity:
250 Discs
Unit Size:
Pack of 250
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility discs.
Recommended Applications, HealthcareAntibiotic Code, TILDescription, Oxoid Tilmicosin Antimicrobial Susceptibility discsPackaging, 5 Cartridges/Pk.Quantity, 50 discs/CartridgeConcentration, 15μgUnit Size, Each
Specifications
Specifications
| Antimicrobial Agents | Tilmicosin |
| Certifications/Compliance | ARTG |
| Content And Storage | 2°C to 8°C |
| Country of Origin | UK |
| Antibiotic Code | TIL |
| Product Type | Antimicrobial Susceptibility Discs |
| Test Type | Antimicrobial Susceptibility |
| For Use With (Application) | Manually Determine The Antibiotic Susceptibility of Microorganisms Using Antimicrobial Susceptibility Testing (AST) Methods |
| No. per Pack | 5 x 50 Discs |
| Packaging Type | Box |
| Show More |