missing translation for 'onlineSavingsMsg'
Learn More

Tiglic Acid Benzyl Ester, TRC

Product Code. 30449119
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449119 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449119 Supplier Toronto Research Chemicals Supplier No. TRCT44005550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tiglic Acid Benzyl Ester
CAS 37526-88-8
Molecular Formula C12H14O2
Quantity 50 g
Synonym (2E)-2-Methyl-2-butenoic Acid Phenylmethyl Ester, Benzyl Tiglate, NSC 69098
InChI Formula InChI=1S/C12H14O2/c1-3-10(2)12(13)14-9-11-7-5-4-6-8-11/h3-8H,9H2,1-2H3/b10-3+
SMILES C\C=C(/C)\C(=O)OCc1ccccc1
IUPAC Name benzyl (E)-2-methylbut-2-enoate
Molecular Weight (g/mol) 190.24
Formula Weight 190.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.