missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tiagabine hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GABA uptake inhibitor (IC50 = 67 nM in vivo). Exhibits high affinity and selectivity for the GAT-1 GABA transporter. Anticonvulsant; also attenuates established dynorphin-induced allodynia in a mouse model after systemic administration.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 412.01 |
| Purity | >99% |
| Molecular Weight (g/mol) | 412.00 |
| Chemical Name or Material | Tiagabine hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction