missing translation for 'onlineSavingsMsg'
Learn More

Thymidine, MP Biomedicals™

Product Code. 11482051 Shop All MP Biomedical Products
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g

Product Code. 11482051

Brand: MP Biomedicals 0219475410

Please to purchase this item. Need a web account? Register with us today!

Used in forming hybrid cell lines where cells are incubated in HAT medium. Thymidine, MP Biomedicals, a cell culture reagent, is a nucleoside composed of deoxyribose (a pentose sugar) joined to the pyrimidine base thymine. It is highly stable and appears

Mainly used in forming hybrid cell lines using HAT (hypoxanthine-aminopterin-thymidine) medium to give rise to monoclonal antibodies. Also used in HT and HAT cocktails for hybridoma fusion, selection and cloning.

Deoxythymidine can be phosphorylated with one, two or three phosphoric acid groups, creating respectively dTMP, dTDP or dTTP (deoxythymidine mono-, di-, or triphosphate). The stability of deoxythymidine under standard temperature and pressure (STP) is very high. Thymidine is listed as a chemical teratogen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Melting Point 185°C to 191°C
Quantity 10 g
Solubility Information Soluble in water, methanol, hot alcohol, hot acetone, hot ethyl acetate pyridine, glacial acetic acid and HCl (20mg/mL, clear, colorless solution); sparingly soluble in hot chloroform.
Optical Rotation 17.5–19.5° (c=1, water)
Formula Weight 242.2
Percent Purity >99.9%
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.