Learn More
Thulium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™
Product Code. 11324128
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. 11324128
Brand: Thermo Scientific Alfa Aesar 041422.09
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13760-79-7
Molecular Formula F3Tm
Molecular Weight (g/mol) 225.93
MDL Number MFCD00049611
InChI Key AATUHDXSJTXIHB-UHFFFAOYSA-K
IUPAC Name thulium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tm+3]
Specifications
Melting Point 1158°C
Density 7.9
Boiling Point 2200°C
Packaging Plastic bottle
Quantity 10 g
UN Number UN3288
Sensitivity Hygroscopic
Specific Gravity 7.9
Formula Weight 225.93
Percent Purity 99.9%
Physical Form Pieces
Chemical Name or Material Thulium(III) fluoride