Learn More
Terumo™ Three-Part Syringe
Product Code. 17122724
Click to view available options
Needle Gauge:
21 G
Unit Size:
Pack of 100
Product Code. 17122724
Brand: Terumo™ 8MDSS10S2138E
Edge
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Three-Part Syringes feature a unique double beveled design for greater patient comfort.
  • Ultra-sharp needles
  • Clear barrels
  • Bold, precise scale markings
  • Smooth plunger action
  • Tamper evident peel packs
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Polypropylene
Needle Gauge 21 G
Latex Free Yes
Length (Metric) 40 mm
Product Type 3-Part Syringe
Volume (Metric) 10 mL