Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key threaded, SBW -J glass
Product Code. 15921511 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
12.5 NS
14.4 NS
18.8 NS
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
15921511 14.4 NS Pack of 100
10384951 12.5 NS Pack of 100
10568031 18.8 NS Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15921511 Supplier DWK Life Sciences Supplier No. 286039206
  • ONLINE EXCLUSIVE PRICE
    85.64€
    71.52€ / Pack of 100
    Save 14.12€
    16% Off
Estimated Shipment: 29-12-2026
To secure the tap plug in the ground DURAN™ single way stopcock.
The 3-part tap safety device holds and secures the threaded tap plug in the tap sleeve.
  • 3-part, complete for SBW-J tap plug
  • Greater safety through installation of the 3-part screw locking device
Specifications
Material Polypropylene
Autoclavable Not Autoclavable
Closure Size 14.4 NS
No. per Pack 100/Pk.
For Use With (Application) For securing the threaded tap plug in the ground DURAN™ one-way tap