Learn More
SCAT™ PTFE Thread Adapters
Shop All SCAT Europe Products
Click to view available options
Closure Size:
GL/S40 (f) to GL45 (m)
GL/S40 (f) to S55 (m)
GL32 (f) to GL45 (m)
GL38 (f) to GL45 (m)
GL45 (f) to S55 (m)
GL45 (m) to 2 in. Tri-sure (m)
GL45 (m) to NS29/32 mm (m)
GL45 (m) to R2 in. BSP/G2 in. (m)
GPI 38-23 (f) to GL45 (m)
ND9
S51 (f) to S55 (m)
S60/61 (m) to B83 (f)
UNF 1/4 in. 28 G (f) to GL14 (m)
Unit Size:
Each
Description
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polytetrafluoroethylene (PTFE) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
| Closure Size | GL45 (m) to NS29/32 mm (m) |
| For Use With (Equipment) | Ground Neck Bottle |
| Material | PTFE |
| Product Type | Thread Adapter |