Learn More
Thiothiamine, TRC
Product Code. 30009680
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009680 100 mg 100mg
17920605 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009680 Supplier Toronto Research Chemicals Supplier No. TRCT375250100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 299-35-4
Molecular Formula C12 H16 N4 O S2
Molecular Weight (g/mol) 296.41
Synonym 2(3H)-Thiazolethione, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, 4-Thiazoline-2-thione, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl- (8CI), 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-2(3H)-thiazolethione, SB, Thiamin thiothiazolone, Thiothiamin, Thiothiamine, 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methylthiazol-2(3H)-thione, Thiamine Hydrochloride Imp. E (EP), Thioxothiamine
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazole-2-thione
SMILES CC1=C(CCO)SC(=S)N1Cc2cnc(C)nc2N
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C12H16N4OS2/c1-7-10(3-4-17)19-12(18)16(7)6-9-5-14-8(2)15-11(9)13/h5,17H,3-4,6H2,1-2H3,(H2,13,14,15)
Formula Weight 296.0766
Chemical Name or Material Thiothiamine