Learn More
Epredia™ Thinline Slotted Cassettes in Tube Packs
Product Code. 16324106 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
16324106 Lavender Pack of 750
16384096 Beige Pack of 750
16394096 Blue Pack of 750
16304106 Green Pack of 750
16314106 Gray Pack of 750
16334106 Light Green Pack of 750
16344106 Lilac Pack of 750
16354106 Orange Pack of 750
16364106 Pink Pack of 750
16374106 Tan Pack of 750
16384106 White Pack of 750
16394106 Yellow Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 16324106 Supplier Epredia™ Supplier No. B851736LA
  • ONLINE EXCLUSIVE PRICE
    158.00€
    122.75€ / Pack of 750
    Save 35.25€
    22% Off
 Restricted Item
In Stock
Add to basket
Add to basket
For routine tissue samples, cassettes are compatible with all Epredia PrintMate™ Cassette Printers and have a front lid hinge with color-matched, attached lids available in 12 different color varieties.
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slots
Closure Included Yes
Quantity 750/Cs.
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color Lavender
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Slotted Cassette