Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Click to view available options
Pore Size:
0.4 μm
1.0 μm
3 μm
3.0 μm
8.0 μm
No. of Wells:
6
12
24
Pore Density:
0.15 x 106 pores/cm2
0.6 x 106 pores/cm2
1 x 108 pores/cm2
2 x 106 pores/cm2
Unit Size:
Pack of 24
Pack of 48
Description
- Clear polystyrene housing with sealed PET capillary pore membrane
- Hanging geometry provides protection to lower compartment cell layer
- Simplified pipetting due to self-lift geometry
- Eccentric in-well positioning allows unequalled access to lower compartment
- Available in 6, 12 and 24 well format
Specifications
Specifications
| Culture Area | 452.4 mm2 |
| Material | Polystyrene |
| Volume (Metric) Working | 2.0 to 4.15 mL |
| Pore Density | 2 x 106 pores/cm2 |
| Pore Size | 3.0 μm |
| Sterility | Sterile |
| No. of Wells | 6 |