Learn More
Thieno[3,2-c]pyridine 4-Methylbenzenesulfonate, TRC
Product Code. 30456858
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456858 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456858 Supplier Toronto Research Chemicals Supplier No. TRCT344515500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Thieno[3,2-c]pyridine 4-Methylbenzenesulfonate
CAS 153075-10-6
Molecular Formula C28 H40 O7
Quantity 500 mg
InChI Formula InChI=1S/C28H40O7/c1-5-7-24(33)35-28(22(31)16-34-23(32)6-2)13-11-20-19-9-8-17-14-18(29)10-12-26(17,3)25(19)21(30)15-27(20,28)4/h14,19-21,25,30H,5-13,15-16H2,1-4H3/t19-,20+,21-,25-,26-,27-,28?/m0/s1
SMILES CCCC(=O)OC1(CC[C@@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)CC
IUPAC Name [(8S,9R,10R,11S,13S,14R)-11-hydroxy-10,13-dimethyl-3-oxo-17-(2-propanoyloxyacetyl)-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate
Molecular Weight (g/mol) 488.613
Formula Weight 488.277