missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thidiazuron

Product Code. 15448109
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448109 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15448109 Supplier Thermo Scientific Chemicals Supplier No. J66663.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

220.25 g/mol

Thidiazuron is a plant growth regulator used in micropropagations to supplement media such as Murashige and Skoog medium. TDZ promotes plant organogenesis and plant regeneration. Thidiazuron enables the production of virus-free stock plants, speeds up breeding, and provides new genotypes for the market.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 220.25
Quantity 250 mg
Packaging Plastic bottle
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.