missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Thiazovivin
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
Description
- Thiazovivin is a selective inhibitor of Rho-associated coiled-coil containing protein kinase (ROCK), a serine/threonine kinase that plays a role in cell polarity, contraction, and actin cytoskeleton reorganization (Xu et al.) Thiazovivin is effective at 5-fold-lower concentrations than another common ROCK inhibitor Y-27632 (Catalog #72302; Xu et al.)
- Promotes survival of human embryonic stem (ES) cells during dissociation by stabilizing E-cadherin and improves cell attachment (Xu et al.)
- Promotes survival of single human induced pluripotent stem (iPS) cells during transfection for TALEN-mediated genome editing (Sun and Zhao)
- Increases the efficiency of reprogramming human somatic cells to iPS cells, in combination with PD0325091 and SB431542 (Lin et al.)
- Increases the efficiency of reprogramming human cord blood mononuclear cells to iPS cells (Hu et al.)
Specifications
Specifications
| CAS | 1226056-71-8 |
| Synonym | Tzv |
| Quantity | 5 mg |
| Molecular Formula | C15H13N5OS |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction