Learn More
Corning™ Thermowell™ Gold 96-Well Polypropylene PCR Microplates
Product Code. 10777724 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Skirt Style:
Elevated
Full
Half
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Unit Size
10777724 Clear Full Pack of 50
10591013 Clear Half Pack of 50
10468102 Clear Elevated Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10777724 Supplier Corning™ Supplier No. 3752
  • ONLINE EXCLUSIVE PRICE
    500.00€
    467.19€ / Pack of 50
    Save 32.81€
    6% Off
Estimated Shipment: 04-02-2027
Available in full, half, and elevated skirt styles
Corning Thermowell GOLD 96-well PCR microplates are offered in four formats to ensure maximum flexibility and a perfect match for your applications. The original Thermowell 96-well PCR microplates are universal fit and can be cut into 3 x 8 well segments.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DNase, RNase-free
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Skirt Style Full
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) For automation, RT-PCR applications
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Well Color Clear