Learn More
Sonoco™ ThermoSafe PolarPack™ Standard
Product Code. 17663585
Change view
Click to view available options
Dimensions (LxWxH):
10.5 x 7.5 x 1.25 in.
6 x 5.5 x 1 in.
6.5 x 5.5 x 1 in.
8 x 5.5 x 1.25 in.
Weight (English):
12 oz.
16 oz.
20 oz.
48 oz.
Material:
Polyethylene
Polyethylene Film
Unit Size:
Pack of 12
Pack of 24
Pack of 36
Pack of 48
Pack of 768
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxWxH) Weight (English) Material Unit Size
17663585 8 x 5.5 x 1.25 in. 20 oz. Polyethylene Pack of 24
13089332 6 x 5.5 x 1 in. 12 oz. Polyethylene Pack of 48
13099332 6.5 x 5.5 x 1 in. 16 oz. Polyethylene Film Pack of 36
16383356 8 x 5.5 x 1.25 in. 20 oz. Polyethylene Pack of 768
13029342 10.5 x 7.5 x 1.25 in. 48 oz. Polyethylene Film Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17663585 Supplier Sonoco™ Supplier No. PP20IN24BL
  • ONLINE EXCLUSIVE PRICE
    93.93€
    52.40€ / Pack of 24
    Save 41.53€
    44% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-12-2026
PolarPack refrigerants provide thermal solutions for temperature critical packages during transit. Available in a broad selection of styles and a multitude of sizes, our refrigerants are designed to provide reliable temperature protection for a vast array of temperature-sensitive products and packages.
  • Thermal solutions for temperature critical packages during transit
  • Gel will phase change at 0° C
  • Condition coolant in freezer before shipments
  • Refrigeration for transport of food, pharmaceuticals and medical products
  • For frozen or refrigerated shipments
  • Alternative to wet ice
  • Quantity: Pack of 768 pieces
TRUSTED_SUSTAINABILITY
Specifications
Quantity 768/Pk.
Weight (English) 20 oz.
Dimensions (LxWxH) 8 x 5.5 x 1.25 in.
Material Polyethylene
Freezing Point 0°C
For Use With (Application) For shipping temperature critical packages during transit