Learn More
Brannan™ Brass Sugar and Jam Thermometer
Click to view available options
Temperature Range (Metric):
40°C to 200°C
Unit Size:
Each
Description
- Brass construction
- Black temperature scale
- Recommended cooking temperature labels
Specifications
Specifications
| Includes | Recommended cooking temperature zone labels |
| Material | Brass |
| Thermometer Fill | Red Spirit |
| Temperature Range (Metric) | 40°C to 200°C |