Learn More
VEE GEE Thermometer, Encapsulated in FEP
Product Code. 17881214 Shop All Heathrow Scientific Products
Change view
Click to view available options
Temperature Range (English):
-30 to +120°F
-31 to +122°F
-4 to +230°F
-4 to +302°F
0 to 300°F
30 to 220°F
Subdivision:
1°C (1.8°F)
1.1°C (2°F)
Immersion Length:
30.4 cm (12 in.)
30.48 cm (12 in.)
39 cm (15.4 in.)
To immersion line
Immersion Type:
Partial Immersion
Total Immersion
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (English) Subdivision Immersion Length Immersion Type unitSize
17881214 -4 to +302°F 1°C (1.8°F) To immersion line Partial Immersion Each
17871254 -30 to +120°F 1.1°C (2°F) 30.48 cm (12 in.) Total Immersion Each
17851194 -4 to +230°F 1.1°C (2°F) 30.48 cm (12 in.) Total Immersion Each
17811244 0 to 300°F 1.1°C (2°F) 30.48 cm (12 in.) Total Immersion Each
17831234 30 to 220°F 1.1°C (2°F) 39 cm (15.4 in.) Total Immersion Each
17881244 -31 to +122°F 1°C (1.8°F) 30.48 cm (12 in.) Total Immersion Each
17841244 -4 to +230°F 1°C (1.8°F) 30.4 cm (12 in.) Total Immersion Each
17881204 -4 to +302°F 1°C (1.8°F) 30.48 cm (12 in.) Total Immersion Each
17801234 -30 to +120°F 1.1°C (2°F) To immersion line Partial Immersion Each
17831204 -4 to +230°F 1.1°C (2°F) To immersion line Partial Immersion Each
17811214 0 to 300°F 1.1°C (2°F) To immersion line Partial Immersion Each
17841214 30 to 220°F 1.1°C (2°F) To immersion line Partial Immersion Each
17871234 -31 to +122°F 1°C (1.8°F) To immersion line Partial Immersion Each
17851214 -4 to +230°F 1°C (1.8°F) To immersion line Partial Immersion Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17881214 Supplier VEE GEE Supplier No. 80908E
The encapsulated glass thermometer is designed to safeguard against broken glass and liquid. It contains a biodegradable Safety Blue fill, offering mercury-like properties minus the associated risks of contamination and hazards.
  • FEP encapsulation- in the event of breakage the FEP coating contains the broken glass and liquid, preventing injury or contamination
  • Feature Safety Blue fill against a white background for easy reading
  • Accuracy is equal to one subdivision
  • Immersion: Total or Partial
Specifications
Bottle Fill Safety Blue
For Use With (Application) Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications
Immersion Type Partial Immersion
Length (Metric) 30.48 cm
Material Glass
Probe Type Immersion/General Purpose (Tapered)
Thermometer Fill Safety Blue
Weight (Metric) 5 g
Product Type Serialized Glass Thermometer
Max. Temperature (English) 302°F
Max. Temperature (Metric) 150°C
Min. Temperature (English) -4°F
Min. Temperature (Metric) -20°C
Temperature Range (English) -4 to +302°F
Temperature Range (Metric) -20 to 150°C
Accuracy ±1 subdivision
Subdivision 1°C (1.8°F)
Immersion Length To immersion line
Width (Metric) 1.5 cm