Learn More
Brannan™ Classroom Kit Thermometer
Brannan™ Classroom Kit Thermometer comes in a blue plastic tray and includes a variety of laboratory thermometers and a set of classroom notes.
Brand:  Brannan™ 25/700/0
Additional Details : Weight : 1.00000kg
Product Code. 13008573
  • 73.90€ / 1 set
please call +351214253350
Add to basket
Description
Description
  • Blue plastic tray that fits into standard classroom storage systems.
  • Includes a variety of laboratory thermometers and a set of classroom notes.
  • Developed for use in the classroom and designed to assist teachers in fulfilling the national curriculum.
  • All thermometers supplied are filled with green spirit.
Specifications
Specifications
Blue plastic case, 8 x 150mm initial™ thermometers; 7 x 300mm initial™ thermometers; 1 x 300mm uncalibrated thermometer and detailed classroom notes
Thermometer kit