Learn More
Amsta Rostfritt Stainless Steel Thermo Flask With Tap and High Skirtband For Serving
Click to view available options
Capacity (Metric):
3 L
5 L
10 L
Unit Size:
Pack of 10
Description
- Stainless steel 304 /1.4301
Specifications
Specifications
| Product Type | Pail |
| Capacity (Metric) | 3 L |
| Color | Silver |
| Material | Stainless Steel |