Learn More
Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple
Product Code. 11706624 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (English):
120 in. (probe)
60 in. (probe)
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
11706624 60 in. (probe) Pack of 6
11841280 120 in. (probe) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11706624 Supplier Cole-Parmer™ Supplier No. 0850585
  • ONLINE EXCLUSIVE PRICE
    157.22€
    116.36€ / Pack of 6
    Save 40.86€
    25% Off
Estimated Shipment: 28-12-2026
Use these probes for a variety of high-temperature applications. Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple Probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Flexible Kapton-insulated wire is ideal for checking food temperatures
  • Flexible fiberglass-Insulated wire used for high-temperature measurements
  • Custom designs or changes to this product are available upon request
  • Exposed-junction probes are not recommended for use with aqueous solutions
  • Do not subject type J exposed-junction probes to moisture as they will rust
Specifications
Type Thermocouple Probe
For Use With (Application) Ideal for Checking Food Temperatures
Insulation Kapton
Length (English) 60 in. (probe)