Learn More
Thermo Scientific™ TX-100/100S Clinical Swinging Bucket Rotor Adapters
Adapters for TX-100S Clinical Rotor and TX-100 Clinical Rotor
21.05€ - 45.85€
Specifications
Type | Rotor Adapter |
---|---|
For Use With (Equipment) | TX-100 and TX-100S clinical swing bucket rotor |
Product Code | Brand | Tube Type | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Type | Description | Price | Quantity & Availability | ||||
11611750
|
Thermo Scientific™
11172287 |
3 mL Blood Collection Tube | 3mL blood tube adapter for centrifuge swinging bucket rotor |
21.05€
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||
11621750
|
Thermo Scientific™
11172288 |
Microtube | 1.5/2 mL microtube adapter for centrifuge swinging bucket rotor |
45.85€
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||
Description
Adapters for TX-100S Clinical Rotor and TX-100 Clinical Rotor
Specifications
Rotor Adapter | |
TX-100 and TX-100S clinical swing bucket rotor |