man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Centrifuges
Centrifuge Adapters
Thermo Scientific™ Sorvall™ Racks for Drum Rotor
Print
Thermo Scientific™ Sorvall™ Racks for Drum Rotor
26.10€
See promotional offers below
Specifications
For Use With (Equipment)
Drum rotor
No. of Places
10
View More Specs
Products
1
Documents
Product Certifications
Products
1
Product Code
Brand
Color
No. of Places
Tube Size
Price
Quantity & Availability
Product Code
Brand
Color
No. of Places
Tube Size
Price
Quantity & Availability
10612612
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Thermo Scientific™
76001244
Red
10
1.5/2mL
26.10€
Each
View alternative products
Save Up to
»
Estimated Shipment: 07-06-2024
Log in
Specifications
For Use With (Equipment)
Drum rotor
No. of Places
10
Documents
Product Certifications
Back to Top