Learn More
Thermo Scientific™ Sorvall™ Legend™ Centrifuge Carts
Product Code. p-317833 Shop All Thermo Scientific Products
Click to view available options
Width (Metric):
55.1 cm
74.9 cm
Height (Metric):
29.0 cm
59.9 cm
Unit Size:
Each
Product Code. 10381712
Brand: Thermo Scientific™ 76000076
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Allow you to operate your centrifuge operation away from, or under, your primary workbench
  • For all Legend T Series centrifuges; sizes as shown in the table
  • Available in bench-high or under-cabinet models
  • With casters
TRUSTED_SUSTAINABILITY
Specifications
Type Mobile Cart
For Use With (Equipment) Thermo Scientific™ Sorvall™ Legend™ RT, Mach 1.6R
Length (Metric) 66.5 cm
Height (Metric) 29.0 cm
Width (Metric) 74.9 cm