Learn More
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
Product Code. 10568761 Shop All Thermo Scientific Products
Click to view available options
Color:
Brown
Gray
Green
Green/Yellow
Red
White
Unit Size:
Each
Product Code. 10568761
Brand: Thermo Scientific™ 75008188
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172
  • Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 4 x 180 Swing bucket rotor
Color Gray