Learn More
Epredia™ Shandon™ Tissue-Loc™ Tissue Cassettes IV
Product Code. 12570077 Shop All Epredia Products
Click to view available options
Color:
Blue
Lilac
White
Yellow
Unit Size:
Pack of 250
Product Code. 12570077
Brand: Epredia™ 41722
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Compatible with Epredia™ Shandon™ MicroWriter™ and PrintMate™ cassette labelers
  • Disposable
  • Holds tissue specimens during embedding, processing
  • Pore style: Small slots
  • Supplied in box of 250 cassettes with attached lids
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Slots
Closure Type Hinged
Color Yellow