Learn More
Epredia™ Cassette 1 Biopsy Cassettes
Product Code. 12790918 Shop All Epredia Products
Click to view available options
Color:
Pink
White
Unit Size:
Pack of 500
Product Code. 12790918
Brand: Epredia™ 1001090
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specially designed to hold biopsy specimens during processing and embedding
  • Made of acetal polymer
  • Cassette writing face is at a 30° angle
  • Pore style is small square
  • Locking lid attached by two hinges
  • Supplied in a box of 500 cassettes with attached breakaway lids
Order Info
Not available in Tube Packs
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Square
Writing Face Angle 30°
Closure Type Hinged
Color Pink
Material Acetal Polymer
Product Type Biopsy Cassette