Learn More
Thermo Scientific™ Rheodyne 9010 Sample Injector
Product Code. 10594855 Shop All Thermo Scientific Products
Click to view available options
Material:
PEEK
Unit Size:
Each
Product Code. 10594855
Brand: Thermo Scientific™ 9010
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
20μL sample loop
Single-mode sample injector designed for the complete filling method
Single-mode sample injector designed for the complete filling method.
Includes: 20μL sample loop
  • Compatible with sample loop sizes 5μL to 10mL
  • PEEK stator
  • Position sensing switch provides a reproducible start signal
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) Sample 20 μL
For Use With (Equipment) Sample Loops 5 to 50 μL
Material PEEK
Type Sample Injector
No. of Ports 6 Ports
Stator Material PEEK