Learn More
Thermo Scientific™ Remel™ Bacillus cereus ATCC™ 14579™, Culti-Loops™
Product Code. 12998848
Click to view available options
:
Bacillus cereus ATCC 14579
Product Code. 12998848
Brand: Thermo Scientific™ R4601217
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
Certifications
Culti-Loops are CE marked.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 14579™*
Product Type Culti-Loop
Strain Designations WDCM00034, [BCRC 10603, CCM 2010, CCUG 7414, CIP 66.24, DSM 31, HAMBI 1887, HAMBI 1905, IAM 12605, JCM 2152, LMG 6923, NBRC 15305, NCCB 75008, NCIMB 9373, NCTC 2599, NRRL B-3711, VKM B-504]
Species Bacillus cereus
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.