missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Premium Grade SPDP
Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Thermo Scientific Pierce Premium Grade SPDP is our highest quality formulation of this pyridyldithiol-based amine-to-sulfhydryl crosslinker, specially characterized for applications where product integrity and risk minimization are paramount. SPDP is a disulfide crosslinker used for reversible (reducible) amine-to-sulfhydryl conjugation.
- High quality - identity and purity confirmed by multiple tests, including quantitative NMR
- Product integrity - enhanced level of testing and characterization compared to standard grade
- Lot retention - ample supply of past lots retained to help ensure future process testing
- Change management - Change Control Notification (CCN) service
- Consistent manufacture - batch-specific manufacturing documentation review
SPDP is N-succinimidyl 3-(2-pyridyldithio)-propionate, a crosslinker for amine-to-sulfhydryl conjugation that forms cleavable (reducible) disulfide bonds with cysteine sulfhydryls. At one end is an N-hydroxysuccinimide (NHS) ester that will react with a primary amine to form a stable amide bond. At the other end is a pyridyl disulfide group that will react with a sulfhydryl group to form a reversible disulfide bond.
Specifications
Specifications
| Chemical Reactivity | Amine-Sulfhydryl |
| Cleavable | By Thiols |
| Molecular Weight (g/mol) | 312.36 |
| PEGylated | No |
| Spacer Arm Length | 6.8 Å |
| Content And Storage | Upon receipt store at -20°C protected from moisture. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Labeling Method | Chemical Labeling |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction