Learn More
Thermo Scientific™ Owl™ Accessories for Giraffe™ Adjustable Gel System
Product Code. 12464687
Click to view available options
Thickness (Metric):
0.08 cm
0.32 cm
Width (Metric):
0.1 cm
16 cm
Type:
Blank Glass Plate
Notched Glass Plate
Spacer Set
Unit Size:
Each
Product Code. 12464687
Brand: Thermo Scientific™ P516R
Glass and spacer set
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ADJ1 Giraffe Adjustable Gel Electrophoresis System
Type Notched Glass Plate
Material Glass
Thickness (English) 0.12 in.
Thickness (Metric) 0.32 cm
Length (Metric) 16 cm
Width (Metric) 16 cm